2-Thiophenemethanethiol 98%

Warning

Synonyms: Thenyl Mercaptan2-(Mercaptomethyl)thiophene2-Thenylmercaptan

AC26511-0010
200022-932EA 178 USD
200022-932
2-Thiophenemethanethiol 98%
2-Thiophenemethanethiol
Formula: C₅H₆S₂
MW: 130.22 g/mol
Boiling Pt: 90 °C (15 mmHg)
Density: 1.2 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00107135
CAS Number: 6258-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR