(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%

Danger

Synonyms: (+)-Mosher's Acid Chloride(S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride(+)-MTPA-Cl

AC26411-2500 AC26411-0010
103527-188EA 143 USD
103527-188 200008-262
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Formula: C₁₀H₈ClF₃O₂
MW: 252.62 g/mol
Boiling Pt: 213…214 °C (760 mmHg)
Density: 1.34 g/cm³
Flash Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00067105
CAS Number: 20445-33-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR