(±)-Tetrahydro-2-furoic acid 98+%

Danger

Synonyms: 2-Tetrahydrofuran Carboxylic AcidTetrahydrofuran-2-carboxylic acid

AC26314-5000 AC26314-0250 AC26314-1000
103532-014EA 38.8 USD
103532-014 200047-098 200022-930
(±)-Tetrahydro-2-furoic acid 98+%
(±)-Tetrahydro-2-furoic acid
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 128…129 °C (13 mmHg)
Density: 1.17 g/cm³
Flash Pt: 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00022473
CAS Number: 16874-33-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR