Hydroxychloroquine sulfate 98%

Warning

Synonyms: 2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol sulfate7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline SulphateN4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine sulphate2-((4-((7-Chloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol sulphateN4-(7-Chloro-4-quinolyl)-N1-ethyl-N1-(2-hydroxyethyl)-1,4-pentanediamine sulfate7-Chloro-4-[4-[N-ethyl-N-(2-hydroxyethyl)amino]-1-methylbutylamino]quinoline sulfate

AC26301-0250 AC26301-1000
103511-934EA 522 USD
103511-934 200012-708
Hydroxychloroquine sulfate 98%
Hydroxychloroquine sulfate
Formula: C₁₈H₂₆ClN₃O
MW: 433.95 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00078203
CAS Number: 747-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR