Glycerol formal (=1,3-dioxan-5-ol) mixture of isomers 99+% stabilized

Warning

Synonyms: 5-Hydroxy-1,3-dioxane1,3-Dioxan-5-ol

AC25977-0050 AC25977-2500 AC25977-5000
200031-472EA 25 USD
200031-472 200012-598 200012-596
Glycerol formal (=1,3-dioxan-5-ol) mixture of isomers 99+% stabilized
Glycerol formal (=1,3-dioxan-5-ol)

Appearance: Clear liquid

Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 193…195°C (760 mmHg)
Density: 1.21 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD10039829
CAS Number: 4740-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR