N-Methylhippuric acid 99%

Warning

AC25976-0250 AC25976-0050
200026-208EA 271.1 USD
200026-208 200020-114
N-Methylhippuric acid 99%
N-Methylhippuric acid

Appearance: White Crystalline powder

Formula: C₁₀H₁₁NO₃
MW: 193.202 g/mol
Melting Pt: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00144940
CAS Number: 2568-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR