Dimethyl (+)-2,3-O-isopropylidene-D-tartrate 97%

Synonyms: (4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic Acid Dimethyl EsterDimethyl (4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylate(+)-2,3-O-Isopropylidene-D-tartaric Acid Dimethyl Ester(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate

AC25844-0100
200033-220EA 119 USD
200033-220
Dimethyl (+)-2,3-O-isopropylidene-D-tartrate 97%
Dimethyl (+)-2,3-O-isopropylidene-D-tartrate
Formula: C₉H₁₄O₆
MW: 218.21 g/mol
Boiling Pt: 139 °C (21hPa)
Density: 1.15 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00040504
CAS Number: 37031-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR