D-(+)-Altrose 99%
AC25802-1000
200024-908EA
249
USD
200024-908
D-(+)-Altrose 99%
D(+)-Altrose
Formula:
C₆H₁₂O₆ Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075620 CAS Number: 1990-29-0 EINECS: 217-870-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...