Butoxyacetic acid 98%
|
Danger
|
Synonyms:
2-Butoxyacetic acid
AC25776-0010
200016-432EA
90.2
USD
200016-432
Butoxyacetic acid 98%
Butoxyacetic acid
Appearance: Clear colorless to light yellow Liquid
Formula:
C₆H₁₂O₃ Boiling Pt: 108 °C Density: 1.01 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00067044 CAS Number: 2516-93-0 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...