3-Chloroperoxybenzoic acid 70-75% in water balance 3-Chlorobenzoic acid and water

Danger

AC25579-1000 AC25579-0010
200056-930EA 96.8 USD
200056-930 200056-928
3-Chloroperoxybenzoic acid 70-75% in water balance 3-Chlorobenzoic acid and water
3-Chloroperoxybenzoic acid
Formula: C₇H₅ClO₃
Melting Pt: 92…94 °C
Density: 0.56 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00002127
CAS Number: 937-14-4
UN: 3106
ADR: 5.2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR