Mercury(II) acetate 99+%

Danger

Synonyms: Acetic acid mercury (II) saltMercury diacetateMercuric acetate

AC25565-1000 AC25565-0010 AC25565-0050
103527-136EA 19.3 USD
103527-136 103527-138 102777-658
Mercury(II) acetate 99+%
Mercury (II) acetate
Formula: (H₃CCOO)₂Hg
MW: 318.68 g/mol
Melting Pt: 179…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS Number: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR