Dichloromethyloctylsilane ≥97%

Danger

Synonyms: OctylmethyldichlorosilaneDichloro(methyl)(octyl)silane

AC25526-0250 AC25526-0050
200044-314EA 24.3 USD
200044-314 103531-992
Dichloromethyloctylsilane ≥97%
Dichloromethyloctylsilane
Formula: C₉H₂₀Cl₂Si
MW: 227.25 g/mol
Boiling Pt: 230 °C
Density: 0.97 g/cm³
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00042606
CAS Number: 14799-93-0
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR