(2R,3R)-(+)-2,3-O-Benzylidene-D-threitol 98%

Synonyms: (+)-2,3-O-Benzylidene-D-threitol

AC25484-0010
200045-206EA 47 USD
200045-206
(2R,3R)-(+)-2,3-O-Benzylidene-D-threitol 98%
(2R,3R)-(+)-2,3-O-Benzylidene-D-threitol
Formula: C₁₁H₁₄O₄
Melting Pt: 69…74 °C
Storage Temperature: Ambient
CAS Number: 58383-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR