(4-Hydroxyphenyl)acetamide 99%

Warning

Synonyms: 4-Hydroxyphenylacetamide

AC25427-0050
200042-054EA 21.02 USD
200042-054
(4-Hydroxyphenyl)acetamide 99%
(4-Hydroxyphenyl)acetamide
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00017145
CAS Number: 17194-82-0
EINECS: 241-235-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR