Sodium-tert-butanolate 99%, pure

Danger

Synonyms: Sodium Tert-ButylateSodium tert-butoxideSodium t-butoxide

AC25255-5000
103527-116EA 171 USD
103527-116
Sodium-tert-butanolate 99%, pure
Sodium tert-butanolate
Formula: C₄H₉NaO
MW: 96.1 g/mol
Melting Pt: 180 °C
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00040575
CAS Number: 865-48-5
EINECS: 212-741-9
UN: 3206
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR