Hyaluronic acid sodium salt 95%

Synonyms: HA sodium saltSodium hyaluronate

AC25177-0050 AC25177-0250 AC25177-0010
103725-484EA 244 USD
103725-484 103725-486 200002-868
Hyaluronic acid sodium salt 95%
Hyaluronic acid sodium salt
Formula: (C₁₄H₂₁NaNO₁₁)n
Storage Temperature: Refrigerator
MDL Number: MFCD00875848
CAS Number: 9067-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR