Theophyllin, anhydrous 99+%

Danger

Synonyms: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione

AC25031-0050 AC25031-0010
200007-858EA 32.3 USD
200007-858 103527-104
Theophyllin, anhydrous 99+%
Theophyllin
Formula: C₇H₈N₄O₂
MW: 180.16 g/mol
Melting Pt: 270…274 °C
Storage Temperature: Ambient
MDL Number: MFCD00079619
CAS Number: 58-55-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR