Isonicotinic anhydride 90% remainder picolinic and nicotinic acid-anhydride

Danger

AC25028-0050
200019-404EA 448 USD
200019-404
Isonicotinic anhydride 90% remainder picolinic and nicotinic acid-anhydride
Isonicotinic anhydride

Appearance: Off-white to beige to light brown Crystalline powder

Formula: C₁₂H₈N₂O₃
Melting Pt: 133 °C
Storage Temperature: Ambient
CAS Number: 7082-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR