Arecolinium bromide 98+%

Danger

Synonyms: Arecoline hydrobromide

AC25013-0050 AC25013-0250 AC25013-1000
200018-188EA 58.51 USD
200018-188 103527-102 200011-260
Arecolinium bromide 98+%
Arecolinium bromide
Formula: C₈H₁₄BrNO₂
MW: 236.11 g/mol
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00039041
CAS Number: 300-08-3
EINECS: 206-087-3
UN: 1544
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR