Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%

Warning

Synonyms: Benzo-18-crown 6-Ether

AC24398-0050
103511-840EA 161 USD
103511-840
Benzo-18-crown-6 (benzo-1,4,7,10,13,16-hexoxacyclooctadecane) 97%
Benzo-18-crown-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecane)
Formula: C₁₆H₂₄O₆
MW: 312.36 g/mol
Melting Pt: 40…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00062741
CAS Number: 14098-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR