Calcium pantothenate 98%

Synonyms: D-Calcium pantothenateCalcium D-PantothenateD-Pantothenic acid calcium saltCalcium D-(+)-pantothenateCalcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoateVitamin B5 calcium salt

AC24330-0050 AC24330-1000 AC24330-5000
200011-542EA 66.1 USD
200011-542 200005-964 103511-832
Calcium pantothenate 98%
Calcium pantothenate
Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00002766
CAS Number: 137-08-6
EINECS: 205-278-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR