Phloroglucide hydrate 95%

Synonyms: PHBF2,3',4,5',6-Pentahydroxybiphenyl hydrate

AC24177-0250
200050-468EA 106 USD
200050-468
Phloroglucide hydrate 95%
Phloroglucide hydrate
Formula: C₁₂H₁₀O₅.xH₂O
MW: 234.207 g/mol (anhy)
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD01543374
CAS Number: 491-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR