3-Hydroxypicolinic acid 98%

Warning

Synonyms: 3-Hydroxypyridine-2-carboxylic acid

AC24105-0250 AC24105-0050
200005-256EA 374 USD
200005-256 103511-818
3-Hydroxypicolinic acid 98%
3-Hydroxypicolinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 208…212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006294
CAS Number: 874-24-8
EINECS: 212-859-0

Order Now

Specification Test Results

Appearance Beige to brown Powder
Infrared spectrum Conforms
Melting point 208 to 214 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR