L(-)-Carnitine 99+%
|
Warning
|
Synonyms:
R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrate
, L-Carnitine
, Levocarnitine
, (R)-3-Hydroxy-4-(trimethylammonio)butanoate
, Vitamin BT
AC24104-0500
AC24104-0010
AC24104-0100
200012-980EA
120
USD
200012-980
103511-814
103511-816
L(-)-Carnitine 99+%
L(-)-Carnitine
Formula:
C₇H₁₅NO₃ MW: 161.20 g/mol Melting Pt: 208…212 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00038747 CAS Number: 541-15-1 EINECS: 208-768-0 ADR: 3,II |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Solubility | (5 % in water) Clear colorless |
Specific optical rotation | -29° to -32° (20 °C, 589 nm) (c=1, H2O) |
Titration with HClO4 | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...