Sesame oil
AC24100-0010
AC24100-2500
200014-404EA
137
USD
200014-404
103467-406
Sesame oil
Sesame oil
Melting Pt:
–5 °C Density: 0.919 g/cm³ Flash Pt: 255 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00132243 CAS Number: 8008-74-0 |
Specification Test Results
Appearance | Clear yellow Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4700 to 1.4760 (20 °C, 589 nm) |
Density | (20 °C) 0.915 to 0.923 |
Acid value | ≤0.6 mg KOH/g |
Peroxide value | ≤10 meq/kg |
Unsaponifiable matter | ≤2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...