Dimethyl-trans-cyclohexane-1,4-dicarboxylate 99+%
|
Warning
|
Synonyms:
Dimethyl trans-1,4-Cyclohexanedicarboxylate
, Dimethyl trans-1,4-cyclohexanedicarboxylate
AC24079-0010
AC24079-0050
200063-518EA
21.7
USD
200063-518
200002-846
Dimethyl-trans-cyclohexane-1,4-dicarboxylate 99+%
Dimethyl trans-cyclohexane-1,4-dicarboxylate
Formula:
C₁₀H₁₆O₄ MW: 200.23 g/mol Melting Pt: 67…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063917 CAS Number: 3399-22-2 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 67 to 70 °C |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...