1,4-Dioxaspiro[4.5]decan-8-one 98%

Warning

Synonyms: 1,4-Cyclohexanedione monoethylene ketal1,4-Cyclohexanedione monoethylene acetal

AC23564-1000 AC23564-0250
103511-798EA 272 USD
103511-798 200002-840
1,4-Dioxaspiro[4.5]decan-8-one 98%
1,4-Dioxaspiro[4.5]decan-8-one
Formula: C₈H₁₂O₃
MW: 156.18 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR