2,6-Diaminopimelic acid 95%

Warning

Synonyms: 1,5-Diaminoheptanedioic Acid

AC23554-0250 AC23554-0050 AC23554-0010
200040-720EA 49.91 USD
200040-720 103511-796 200009-042
2,6-Diaminopimelic acid 95%
2,6-Diaminopimelic acid
Formula: C₇H₁₄N₂O₄
MW: 190.20 g/mol
Melting Pt: 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002637
CAS Number: 583-93-7

Order Now

Specification Test Results

Appearance White to beige to tan Powder and/or chunks
Infrared spectrum Conforms
Titration with HClO4 94.0 to 106.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR