Benzofuran-2-carboxylic acid 98%

Warning

Synonyms: Benzo[b]furan-2-carboxylic acid

AC23326-0010 AC23326-0050
200030-510EA 98.4 USD
200030-510 200024-518
Benzofuran-2-carboxylic acid 98%
Benzofuran-2-carboxylic acid
Formula: C₉H₆O₃
MW: 162.15 g/mol
Melting Pt: 195…196 °C
Storage Temperature: Ambient
EINECS: 207-818-9

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Melting point 192 to 196 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR