Ethyl propionylacetate 98+%

Warning

Synonyms: Ethyl 3-oxopentanoateEthyl 3-oxovalerate

AC23291-0050 AC23291-0250 AC23291-0010
103511-772EA 238 USD
103511-772 103511-770 200012-446
Ethyl propionylacetate 98+%
Ethyl propionylacetate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 83…84 °C (12 mmHg)
Density: 1.01 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00009317
CAS Number: 4949-44-4
EINECS: 225-593-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4240 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR