Azacyclonol 98+%

Warning

Synonyms: alpha,alpha-Diphenyl-4-piperidinemethanolalpha-(4-Piperidyl)benzhydrolDiphenyl(4-piperidinyl)methanolDiphenyl-4-piperidinemethanolα,α-Diphenyl-4-piperidinomethanol

AC23252-0250 AC23252-0050
200011-078EA 26.1 USD
200011-078 200024-358
Azacyclonol 98+%
Azacyclonol
Formula: C₁₈H₂₁NO
MW: 267.37
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00066980
CAS Number: 115-46-8
EINECS: 204-092-5

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 163 °C
Water ≤0.25 % (K.F.)
Loss on drying ≤1 %
Sulfated ash ≤0.25 %
HPLC ≥98.0 %
Appearance of solution (5% soln. in ethanol) clear, colorless
Titration with HClO4 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR