Linalyl acetate (synthetic) 95%

Warning

Synonyms: 3,7-Dimethylocta-1,6-dien-3-yl acetate

AC23230-0050 AC23230-5000 AC23230-1000
200031-702EA 23.2 USD
200031-702 200037-798 200037-800
Linalyl acetate (synthetic) 95%
Linalyl acetate
Formula: C₁₂H₂₀O₂
MW: 196.2860 g/mol
Flash Pt: 85 °C
Storage Temperature: Ambient
EINECS: 204-116-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4520 (20 °C, 589 nm)
GC ≥94 %
Specific gravity (20 °C) 0.898 to 0.903

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR