D-Panthenol 98+%

Synonyms: Dexpanthenol(R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

AC23280-5000 AC23280-1000
200018-990EA 73.6 USD
200018-990 200031-170
D-Panthenol 98+%
D-Panthenol
Formula: C₉H₁₉NO₄
MW: 205.25 g/mol
Boiling Pt: 118…120 °C (2.7Pa)
Storage Temperature: Refrigerator
MDL Number: MFCD00065006
CAS Number: 81-13-0
EINECS: 201-327-3
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid or solid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.5020 (20 °C, 589 nm)
Assay ≥98.0 %
Water ≤1.0 % (K.F.)
Specific optical rotation +29.0° to +32.0° (20 °C, 589 nm) (c=5, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR