γ-Cyclodextrine hydrate 99%

Synonyms: γ-Cyclodextrin xhydrateγ-Cyclodextrin hydrate

AC22990-0050
103511-436EA 889 USD
103511-436
γ-Cyclodextrine hydrate 99%
γ-Cyclodextrine hydrate
Formula: C₄₈H₈₀O₄₀·H₂O
MW: 1297.15 g/mol (anhy)
Melting Pt: 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149574
CAS Number: 91464-90-3
EINECS: 241-482-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤11 %
HPLC ≥98.5 % (ELSD)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR