Citrinin 98%

Danger

Synonyms: Citrinin, yellow(3S,4R)-8-hydroxy-3,4,5-trimethyl-6-oxo-4,6-dihydro-3H-isochromene-7-carboxylic acid

AC22958-0050
200048-838EA 215 USD
200048-838
Citrinin 98%
Citrinin

Appearance: Yellow Crystalline powder or flakes

Formula: C₁₃H₁₄O₅
MW: 250.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006912
CAS Number: 518-75-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or flakes
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR