L-Methionine methyl ester hydrochloride 98%

Synonyms: H-Met-OMe.HClH-Met-OMe·HCl

AC22935-0500 AC22935-0250
200037-814EA 376 USD
200037-814 200031-726
L-Methionine methyl ester hydrochloride 98%
L-Methionine methyl ester hydrochloride
Formula: C₆H₁₃NO₂S·HCl
MW: 199.7 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012491
CAS Number: 2491-18-1
EINECS: 219-651-9

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Solubility (5 % in methanol) Clear colorless to light yellow
Specific optical rotation +24° to +28° (20 °C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR