3-Hydroxybenzylhydrazine dihydrochloride 98%

Warning

AC22903-0050 AC22903-0010
200007-038EA 152 USD
200007-038 200007-036
3-Hydroxybenzylhydrazine dihydrochloride 98%
3-Hydroxybenzylhydrazine dihydrochloride

Appearance: Off-white to beige Adhering powder

Formula: C₇H₁₂Cl₂N₂O
Melting Pt: 138 °C
Storage Temperature: Ambient
CAS Number: 81012-99-9

Order Now

Specification Test Results

Appearance Off-white to beige Adhering powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR