3,4-Dihydroxybenzylamine hydrobromide 98%

Synonyms: 4-(Aminomethyl)benzene-1,2-diol hydrobromide

AC22857-2500 AC22857-0010 AC22857-0050
200047-248EA 293 USD
200047-248 200035-154 200016-852
3,4-Dihydroxybenzylamine hydrobromide 98%
3,4-Dihydroxybenzylamine hydrobromide
Formula: C₇H₁₀BrNO₂
MW: 220.07 g/mol
Melting Pt: >180 °C
Storage Temperature: Ambient
MDL Number: MFCD00012859
CAS Number: 16290-26-9
EINECS: 240-382-8

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR