Pyridoxal-5'-phosphate monohydrate 99%

Synonyms: (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydratePyridoxal 5-phosphate monohydratePyridoxal-5-phosphate hydrate

AC22817-0010 AC22817-0050
200020-552EA 44.3 USD
200020-552 200020-554
Pyridoxal-5'-phosphate monohydrate 99%
Pyridoxal 5'-phosphate monohydrate
Formula: C₈H₁₀NO₆P·1H₂O
MW: 265.17 g/mol (247.15 anhy)
Melting Pt: 140…143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149414
CAS Number: 41468-25-1

Order Now

Specification Test Results

Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Water ≤10 %
HPLC ≥98.5 %
Phosphorus (P) ≤0.2 % (P inorganic)
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR