Methyl-ɑ-D-mannopyranoside 99+%

Synonyms: (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triolɑ-Methyl-D-mannopyranosideɑ-Methyl-D-mannosideMethyl-ɑ-D-Mannoside

AC22925-0250 AC22925-1000
103509-916EA 53.9 USD
103509-916 200011-122
Methyl-ɑ-D-mannopyranoside 99+%
Methyl-alpha-D-mannopyranoside
Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Melting Pt: 186…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00063262
CAS Number: 617-04-9

Order Now

Specification Test Results

Appearance White to off-white to light grey Powder
Infrared spectrum Conforms
Melting point 186 to 196 °C
Water ≤0.5 %
Sulfated ash ≤0.2 %
Solubility (10 % in water) Clear colorless
HPLC ≥99.0 % (ELSD)
Specific optical rotation +77° to +81° (20 °C, 589 nm) (c=10, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR