L(-)-Methioninol 99+%

Warning

Synonyms: (S)-2-Amino-4-(methylthio)butan-1-olL-Methioninol

AC22923-0050 AC22923-0010
103509-914EA 275 USD
103509-914 200019-524
L(-)-Methioninol 99+%
L(-)-Methioninol
Formula: C₅H₁₃NOS
MW: 135.2 g/mol
Melting Pt: 31…33 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004735
CAS Number: 2899-37-8
EINECS: 220-788-1

Order Now

Specification Test Results

Appearance Transparent white Low melting crystalline solid
Infrared spectrum Conforms
GC ≥99.0 %
Specific optical rotation -11.5° to -13.5° (20 °C, 589 nm) (c=1.4, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR