trans-4-(Aminomethyl)cyclohexanecarboxylic acid 98+%

Warning

Synonyms: HAKUTAMCHAAMCAAMCHATranexamic acid

AC22804-2500 AC22804-0500
200002-786EA 160 USD
200002-786 103466-658
trans-4-(Aminomethyl)cyclohexanecarboxylic acid 98+%
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Formula: C₈H₁₅NO₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001466
CAS Number: 1197-18-8
EINECS: 214-818-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
pH 6.5 to 8.0 (5 % solution)
Titration with HClO4 98.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR