4-Bromo-DL-phenylalanine 99%

AC22779-2500 AC22779-0010
200044-360EA 19.2 USD
200044-360 200032-452
4-Bromo-DL-phenylalanine 99%
4-Bromo-DL-phenylalanine
Formula: C₉H₁₀BrNO₂
MW: 244.09
Melting Pt: 262…263 °C
Storage Temperature: Refrigerator
EINECS: 237-941-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR