Thymidine (2'-deoxythymidine) 99+%

Synonyms: dT1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dioneThymine deoxyriboside

AC22674-1000 AC22674-0050
103511-668EA 697 USD
103511-668 200002-736
Thymidine (2'-deoxythymidine) 99+%
Thymidine (2'-Deoxythymidine)
Formula: C₁₀H₁₄N₂O₅
MW: 242.23 g/mol
Melting Pt: 187…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00006537
CAS Number: 50-89-5
EINECS: 200-070-4

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.3 % (800 °C)
HPLC ≥99.0 %
UV spectrophotometry 98.0 to 102.0 % (lambda max. 267 nm, pH 7)
UV spectrophotometry molar absorptivity 9600 +/- 4%, on dry basis
Ratio of Absorbances 0.62 to 0.68 (A250/A260)
Ratio of Absorbances 0.70 to 0.76 (A280/A260)
Specific optical rotation +17.5° to +19.5° (20 °C, 589 nm) (c=3, H2O)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR