2'-Deoxyguanosine hydrate 99+%

Synonyms: Guanine-2′-deoxyriboside9-(2-Deoxy-β-D-ribofuranosyl)guanine

AC22673-0250 AC22673-1000 AC22673-0050 AC22673-0010
200022-614EA 20.9 USD
200022-614 103464-596 200046-816 103464-594
2'-Deoxyguanosine hydrate 99+%
2'-Deoxyguanosine monohydrate
Formula: C₁₀H₁₃N₅O₄·xH₂O
MW: 267.25 g/mol (anhy)
Melting Pt: 300 °C (dec.)
Flash Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150760
CAS Number: 961-07-9
EINECS: 213-505-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤8.0 % (K.F.)
HPLC ≥99.0 %
UV spectrophotometry ≥98.0 % (lambda max. 252.5nm, pH 7)
UV spectrophotometry molar absorptivity 13700 +/- 4%, on dry basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR