2'-Deoxycytidine 99+%

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-onedeoxyC

AC22672-0010 AC22672-0250 AC22672-2500 AC22672-5000
200028-698EA 69.9 USD
200028-698 103464-592 200041-002 200041-004
2'-Deoxycytidine 99+%
2'-Deoxycytidine
Formula: C₉H₁₃N₃O₄
MW: 227.22 g/mol
Melting Pt: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00006547
CAS Number: 951-77-9
EINECS: 213-454-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤8 %
HPLC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR