5-Azacytidine 99%

Danger

Synonyms: 4-Amino-1-(beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-oneLadakamycin

AC22662-0500
200010-680EA 30.4 USD
200010-680
5-Azacytidine 99%
5-Azacytidine
Formula: C₈H₁₂N₄O₅
MW: 244.21 g/mol
Melting Pt: 228…230 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00006539
CAS Number: 320-67-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR