2'-Deoxyinosine ≥98%

Synonyms: 9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

AC22636-1000
200041-008EA 23.4 USD
200041-008
2'-Deoxyinosine ≥98%
2'-Deoxyinosine
Formula: C₁₀H₁₂N₄O₄
MW: 252.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005762
CAS Number: 890-38-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR