Inosine 5'-monophosphate disodium salt hydrate 97%
Synonyms:
Disodium inosinate hydrate
, 5'-Inosinic acid disodium salt hydrate
, Inosic acid disodium salt hydrate
, 5'-IMP 2Na hydrate
, Inosine 5'-monophosphate disodium salt xhydrate
AC22626-0250
AC22626-0050
AC22626-1000
103517-738EA
220
USD
103517-738
200037-654
200019-366
Inosine 5'-monophosphate disodium salt hydrate 97%
Inosine 5'-monophosphate disodium salt hydrate
Formula:
C₁₀H₁₁N₄Na₂O₈P·xH₂O MW: 392.17 g/mol (anhy) Melting Pt: 175 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00150372 CAS Number: 352195-40-5 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤28.5 % (K.F.) |
UV spectrophotometry | ≥96.0 % (lambda max. 249 nm, pH 7) |
UV spectrophotometry | molar absorptivity 12300 +/- 4%, on dry basis |
Heavy metals (as Pb) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...