Guanosine 5'-triphosphate disodium salt hydrate 90% for biochemistry
Synonyms:
GTP disodium salt hydrate
AC22625-2500
AC22625-0500
AC22625-0010
200006-608EA
77.9
USD
200006-608
200006-606
200006-610
Guanosine 5'-triphosphate disodium salt hydrate 90% for biochemistry
Guanosine 5'-triphosphate disodium salt hydrate
Formula:
C₁₀H₁₄N₅Na₂O₁₄P₃·H₂O MW: 567.147 g/mol (anhy) Melting Pt: 180 °C (dec.) Storage Temperature: Freezer |
MDL Number:
MFCD03410297 EINECS: 259-940-7 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤12 % (K.F.) |
HPLC | ≥88 % |
UV spectrophotometry | ≥88 % (lambda max. 252 nm, pH 7) |
Ratio of Absorbances | 1.12 to 1.18 (A250/A260) |
Ratio of Absorbances | 0.63 to 0.69 (A280/A260) |
Related substances | ≤1 % (typically) Guanosine monophosphate (HPLC) |
Related substances | ≤4 % (typically) Guanosine diphosphate (HPLC) |
Sodium (Na) | 7 to 9.5 % (flame photom.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...